accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPbO

CAS RN 1309-60-0

IUPAC Name: plumbanedione
Synonyms: lead dioxide
Molecular Weight (g/mol): 239.2
Molecular Formula: O2Pb
InChi Key: YADSGOSSYOOKMP-UHFFFAOYSA-N
SMILES: O=[Pb]=O
Molecular Weight (g/mol) 
  • (1)
  • (19)
Quantity 
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (4)
  • (2)
  • (8)
  • (1)
Grade 
  • (2)
  • (2)
  • (4)
  • (6)
Physical Form 
  • (6)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (6)
  • (1)
  • (2)
  • (1)
  • (6)
  • (10)
  • (1)
  • (1)
  • (19)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (2)
  • (8)
  • (1)
  • (2)
  • (2)
  • (4)
  • (6)
  • (6)
  • (8)