accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHHNCH3

CAS RN 13093-05-5

IUPAC Name: ethyl[6-(ethylamino)hexyl]amine
Synonyms: ethyl[6-(ethylamino)hexyl]amine
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: LDQWVRMGQLAWMN-UHFFFAOYSA-N
SMILES: CCNCCCCCCNCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results