accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13094-51-4

IUPAC Name: 2-bromoacetyl 2-bromoacetate
Synonyms: 2-bromoacetyl 2-bromoacetate
Molecular Weight (g/mol): 259.88
Molecular Formula: C4H4Br2O3
InChi Key: FUKOTTQGWQVMQB-UHFFFAOYSA-N
SMILES: BrCC(=O)OC(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results