CAS RN 13096-96-3
IUPAC Name:
4-chloro-1H-indazole
Synonyms:
4-chloro-1H-indazole
Molecular Weight (g/mol):
152.58
Molecular Formula:
C7H5ClN2
InChi Key:
CQTGQYVQJOJQCM-UHFFFAOYSA-N
SMILES:
ClC1=C2C=NNC2=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Chloro-1H-indazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.