accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OFFFOFFFFFFOFFF

CAS RN 13098-39-0

IUPAC Name: bis(hexafluoropropan-2-one) trihydrate
Synonyms: bis(hexafluoroacetone) trihydrate
Molecular Weight (g/mol): 386.09
Molecular Formula: C6H6F12O5
InChi Key: VAIZVCMDJPBJCM-UHFFFAOYSA-N
SMILES: O.O.O.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results