accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1309982-16-8

IUPAC Name: 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 237.08
Molecular Formula: C12H17BFNO2
InChi Key: URCVTWMYLLPYQY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(N)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results