accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOSOOOO

CAS RN 131-08-8

IUPAC Name: sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid
Synonyms: silver salt
Molecular Weight (g/mol): 311.26
Molecular Formula: C14H8NaO5S
InChi Key: GGCZERPQGJTIQP-UHFFFAOYSA-N
SMILES: [Na+].OS(=O)(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results