accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C

CAS RN 131-11-3

IUPAC Name: 1,2-dimethyl benzene-1,2-dicarboxylate
Synonyms: dimethyl phthalate dimethyl o-phthalate dimethylphthalate caswell no. 380 unimoll DM palatinol M dmf (insect repellant) DMP NTM solvarone
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: NIQCNGHVCWTJSM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (10)
Boiling Point 
  • (6)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (2)
  • (10)
  • (6)
  • (3)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)