accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131-56-6

IUPAC Name: 4-benzoylbenzene-1,3-diol
Synonyms: benzophenone-1 resbenzophenone uvinul 400 HHB dastib 263 2,4-dihydroxybenzofenon uvistat 12 inhibitor dhbp dhbp cpd eastman inhibitor dhpb
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: ZXDDPOHVAMWLBH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)