accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131-61-3

IUPAC Name: sodium 2,3,4,6-tetrachlorobenzen-1-olate
Synonyms: sodium 2,3,4,6-tetrachlorobenzenolate
Molecular Weight (g/mol): 253.86
Molecular Formula: C6HCl4NaO
InChi Key: YLFFQZKUOUYUFG-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=C(Cl)C(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results