accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeSe

CAS RN 1310-32-3

IUPAC Name: selanylideneiron
Synonyms: iron(II) selenide
Molecular Weight (g/mol): 134.82
Molecular Formula: FeSe
InChi Key: WALCGGIJOOWJIN-UHFFFAOYSA-N
SMILES: [Fe]=[Se]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results