accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHFeFeFeFeOSOOOOSOOOOSOOOOSOOOOSOOO

CAS RN 1310-45-8

IUPAC Name: tetrairon(3+) dihydroxide pentasulfate
Synonyms: ferric subsulfate solution
Molecular Weight (g/mol): 737.67
Molecular Formula: Fe4H2O22S5
InChi Key: GDPKWKCLDUOTMP-UHFFFAOYSA-B
SMILES: [OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (5)
  • (5)
Physical Form 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (10)
  • (2)
  • (8)
  • (3)
  • (5)
  • (5)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)