CAS RN 1310-58-3
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium Hydroxide Solution, 0.5N (N/2) (In Methanol/Certified), Fisher Chemical™
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium Hydroxide (Flakes/Technical), Fisher Chemical
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium Hydroxide Solution, Volumetric, 8.0 M KOH (8.0 N), MilliporeSigma™ Supelco™
High-purity 8.0 M volumetric potassium hydroxide solution for precise analytical applications.
Potassium Hydroxide, 10% (w/v), Ricca Chemical
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, ca. 85%, ACS reagent, pellets
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, pellets, 85%
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, pure, 8N solution in water
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 SMILES: [OH-].[K+]
Potassium hydroxide, 0.1N Standardized Solution in methanol
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium;hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, 99.98%, (trace metal basis), pellets
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, 50% w/v aq. soln.
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium Hydroxide, 0.0100 N (N/100) in Isopropyl Alcohol, Ricca Chemical
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 IUPAC Name: potassium hydroxide SMILES: [OH-].[K+]
Potassium hydroxide, 1N solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
Potassium hydroxide, ca. 85%, extra pure, flakes
CAS: 1310-58-3 Molecular Formula: HKO Molecular Weight (g/mol): 56.11 MDL Number: MFCD00003553 InChI Key: KWYUFKZDYYNOTN-UHFFFAOYSA-M Synonym: potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 PubChem CID: 14797 ChEBI: CHEBI:32035 SMILES: [OH-].[K+]