accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3

CAS RN 13106-53-1

IUPAC Name: 1-hexadecyl-4-methylpyridin-1-ium chloride
Synonyms: 1-hexadecyl-4-methylpyridin-1-ium chloride
Molecular Weight (g/mol): 354.02
Molecular Formula: C22H40ClN
InChi Key: YCMFMSOTJOUSLI-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCC[N+]1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results