accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OSnH

CAS RN 13121-70-5

IUPAC Name: tricyclohexylstannanol
Synonyms: cyhexatin
Molecular Weight (g/mol): 387.2
Molecular Formula: C18H36OSn
InChi Key: ODQJUYRIEATUPA-UHFFFAOYSA-N
SMILES: O.C1CCC(CC1)[SnH](C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results