accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 13121-99-8

IUPAC Name: 2-(pyridin-4-yl)acetonitrile
Synonyms: 2-(pyridin-4-yl)acetonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: BMVSAKPRNWZCPG-UHFFFAOYSA-N
SMILES: N#CCC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results