accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONOOOONOOOONOOOONOORhRh

CAS RN 131219-55-1

IUPAC Name: bis(λ²-rhodium(2+)) tetrakis(2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-phenylpropanoate)
Synonyms: bis(λ²-rhodium(2+)) tetrakis(2-(1,3-dioxoisoindol-2-yl)-3-phenylpropanoate)
Molecular Weight (g/mol): 1382.96
Molecular Formula: C68H48N4O16Rh2
InChi Key: LPPDGVKSVXIYKY-UHFFFAOYNA-J
SMILES: [Rh++].[Rh++].[O-]C(=O)C(CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O.[O-]C(=O)C(CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O.[O-]C(=O)C(CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O.[O-]C(=O)C(CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results