accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 131222-99-6

IUPAC Name: 6,12-dibromochrysene
Synonyms: 6,12-dibromochrysene
Molecular Weight (g/mol): 386.09
Molecular Formula: C18H10Br2
InChi Key: RULVBMDEPWAFIN-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C(Br)C3=CC=CC=C23)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)