accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNNNNCH2FFFSOONSOOFFFFFFSOONSOOFFF

CAS RN 1312310-16-9

IUPAC Name: 1-ethenyl-3-[4-(1-ethenyl-1H-imidazol-3-ium-3-yl)butyl]-1H-imidazol-3-ium; bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: 3-ethenyl-1-[4-(3-ethenylimidazol-1-ium-1-yl)butyl]imidazol-1-ium; bis(bistriflylimide anion)
Molecular Weight (g/mol): 804.61
Molecular Formula: C18H20F12N6O8S4
InChi Key: DHLTUSHCPNNNKG-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C=CN1C=C[N+](CCCC[N+]2=CN(C=C)C=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results