accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHHNClCl

CAS RN 13124-15-7

IUPAC Name: N'-(3,4-dichlorophenyl)ethoxycarbohydrazide
Synonyms: N'-(3,4-dichlorophenyl)ethoxycarbohydrazide
Molecular Weight (g/mol): 249.09
Molecular Formula: C9H10Cl2N2O2
InChi Key: JLVAIPOSBJQZGC-UHFFFAOYSA-N
SMILES: CCOC(=O)NNC1=CC(Cl)=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results