CAS RN 13127-88-3
IUPAC Name:
1-[(²H)oxy](²H₅)benzene
Synonyms:
1-[(²H)oxy](²H₅)benzene
Molecular Weight (g/mol):
100.15
Molecular Formula:
C6H6O
InChi Key:
ISWSIDIOOBJBQZ-QNKSCLMFSA-N
SMILES:
[2H]OC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]
1
–
1
of
1
results
1
Phenol-d6, SPEX CertiPrep™
CAS: 13127-88-3 Molecular Formula: C6H6O Molecular Weight (g/mol): 100.15 MDL Number: MFCD00002144 InChI Key: ISWSIDIOOBJBQZ-QNKSCLMFSA-N PubChem CID: 123295 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-deuteriooxybenzene SMILES: [2H]OC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]