accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131276-24-9

IUPAC Name: 1,8-dibromoanthracene
Synonyms: 1,8-dibromoanthracene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: JQQFVBWHOMXEEA-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=CC3=CC=CC(Br)=C3C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results