CAS RN 1313-30-0
IUPAC Name:
Sodium dodecamolybdophosphate hydrate
Synonyms:
—
Molecular Weight (g/mol):
1891.18
Molecular Formula:
Mo12Na3O40P
InChi Key:
—
SMILES:
*
1
–
1
of
1
results
1
Sigma Aldrich Sulfo-Sda (Sulfo-Nhs-Diazirine) (Sulfosuccinimidyl 4,4'-Azipentanoate)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.