accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSHO

CAS RN 131337-81-0

IUPAC Name: {thieno[2,3-b]pyridin-2-yl}methanol
Synonyms: thieno[2,3-b]pyridin-2-ylmethanol
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: XLUKSWZEEAGBDH-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC=CN=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results