accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCZnI

CAS RN 131379-16-3

IUPAC Name: 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Synonyms: 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Molecular Weight (g/mol): 341.45
Molecular Formula: C9H9IO2Zn
InChi Key: DVAYVJGYZRYJEV-UHFFFAOYSA-M
SMILES: [Zn+]I.CCOC(=O)C1=CC=[C-]C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results