CAS RN 131379-40-3
IUPAC Name:
2-(ethoxycarbonyl)benzen-1-ide; bromozincylium
Synonyms:
2-(ethoxycarbonyl)benzen-1-ide; bromozincylium
Molecular Weight (g/mol):
294.45
Molecular Formula:
C9H9BrO2Zn
InChi Key:
LRSRESIMIGVTLH-UHFFFAOYSA-M
SMILES:
[Zn+]Br.CCOC(=O)C1=CC=CC=[C-]1
1
–
1
of
1
results
1
Sigma Aldrich 3-Hydroxyphenylboronic Acid Pinacol Ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.