accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CP(E)

CAS RN 13138-25-5

IUPAC Name: [(2E)-but-2-en-1-yl]triphenylphosphanium chloride
Synonyms: (2E)-but-2-en-1-yltriphenylphosphanium chloride
Molecular Weight (g/mol): 352.84
Molecular Formula: C22H22ClP
InChi Key: YYTDJYJBYMQMDI-SQQVDAMQSA-M
SMILES: [Cl-].C\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results