accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(S)OHNOHOOCH3CH3CH3

CAS RN 13139-14-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
Synonyms: boc-tryptophan|BOC-TRP-OH
Molecular Weight (g/mol): 304.35
Molecular Formula: C16H20N2O4
InChi Key: NFVNYBJCJGKVQK-ZDUSSCGKSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)