CAS RN 1314-13-2
Zinc oxide, ACS reagent
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, Puratronic™, 99.999% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.9% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, nanopowder, 99% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, ACS, 99.0% min
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.999%, (trace metal basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, NanoArc™ ZN-0605
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc Oxide, 99%, (ACS Reagent Grade), MP Biomedicals
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, NanoTek∣r
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.9% (metals basis), Thermo Scientific™
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]