accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314-15-4

IUPAC Name: dioxoplatinum
Synonyms: adams' catalyst
Molecular Weight (g/mol): 227.08
Molecular Formula: O2Pt
InChi Key: YKIOKAURTKXMSB-UHFFFAOYSA-N
SMILES: O=[Pt]=O
Molecular Weight (g/mol) 
  • (6)
  • (7)
Quantity 
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (5)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (5)
  • (6)
  • (7)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)