accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnOO

CAS RN 1314-22-3

IUPAC Name: zinc(2+) dioxidanediide
Synonyms: zinc peroxide
Molecular Weight (g/mol): 97.38
Molecular Formula: O2Zn
InChi Key: IPTOGCUGCFHDSS-UHFFFAOYSA-N
SMILES: [Zn++].[O-][O-]
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)