CAS RN 1314-23-4
Zirconium(IV) oxide, nanopowder, 99% (metals basis excluding Hf), Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, yttria stabilized, 99.95% (metals basis excluding Hf), Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, Puratronic™, 99.978% (metals basis), Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, 18% in H{2}O, colloidal dispersion, stabilized with 1.3% yttrium oxide, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, yttria stabilized, 99% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium(IV) oxide, cement, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Zirconium oxide, polymeric precursor, Thermo Scientific Chemicals
CAS: 1314-23-4 Molecular Formula: O2Zr Molecular Weight (g/mol): 123.22 MDL Number: MFCD00011310 MFCD01868884 InChI Key: MCMNRKCIXSYSNV-UHFFFAOYSA-N Synonym: zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler PubChem CID: 62395 IUPAC Name: dioxozirconium SMILES: O=[Zr]=O
Sigma Aldrich Zirconium(IV) oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2