accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314-28-9

IUPAC Name: trioxorhenium
Synonyms: rhenium trioxide
Molecular Weight (g/mol): 234.2
Molecular Formula: O3Re
InChi Key: YSZJKUDBYALHQE-UHFFFAOYSA-N
SMILES: O=[Re](=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)