accessibility menu, dialog, popup

UserName

Viewing profile as user:
OTlOTlO

CAS RN 1314-32-5

IUPAC Name: oxo[(oxothallanyl)oxy]thallane
Synonyms: thallium(III) oxide
Molecular Weight (g/mol): 456.76
Molecular Formula: O3Tl2
InChi Key: QTQRFJQXXUPYDI-UHFFFAOYSA-N
SMILES: O=[Tl]O[Tl]=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)