CAS RN 1314-56-3
Phosphorus Pentoxide (Certified ACS), Fisher Chemical™
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphoric Anhydride IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus(V) oxide, 98%
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphorus pentoxide IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus(V) oxide, ACS, 98% min
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus pentoxide, 99+%, for analysis
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphoric anhydride IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus pentoxide, 98+%, ACS reagent
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphoric anhydride IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus pentoxide, 98%, extra pure
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphoric anhydride IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus(V) oxide, 99.99%
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Phosphorus Pentoxide, ACS, 98%, Spectrum™ Chemical
Phosphorus Pentoxide (Laboratory), Fisher Chemical
CAS: 1314-56-3 Molecular Formula: O5P2 Molecular Weight (g/mol): 141.94 MDL Number: MFCD00011440 InChI Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N Synonym: Phosphoric Anhydride IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3