accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314-95-0

IUPAC Name: λ²-tin(2+) sulfanediide
Synonyms: λ²-tin(2+) sulfanediide
Molecular Weight (g/mol): 150.77
Molecular Formula: SSn
InChi Key: DZXKSFDSPBRJPS-UHFFFAOYSA-N
SMILES: [S--].[Sn++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results