accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314-96-1

IUPAC Name: strontium(2+) sulfanediide
Synonyms: strontium(2+) sulfanediide
Molecular Weight (g/mol): 119.68
Molecular Formula: SSr
InChi Key: ZEGFMFQPWDMMEP-UHFFFAOYSA-N
SMILES: [S--].[Sr++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results