accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131418-11-6

IUPAC Name: 2-chloro-N-methylpyridine-4-carboxamide
Synonyms: 2-chloro-N-methylpyridine-4-carboxamide
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: HZCPKEZVYGEGEN-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results