accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314538-55-0

IUPAC Name: potassium tert-butyl N-[(trifluoroboranuidyl)methyl]carbamate
Synonyms: potassium tert-butyl N-[(trifluoroboranuidyl)methyl]carbamate
Molecular Weight (g/mol): 237.07
Molecular Formula: C6H12BF3KNO2
InChi Key: WSOSBCPWIQUNLD-UHFFFAOYSA-N
SMILES: [K+].CC(C)(C)OC(=O)NC[B-](F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results