accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131468-33-2

IUPAC Name: 4,6-dichloro-N-(4-fluorophenyl)-1,3,5-triazin-2-amine
Synonyms: 4,6-dichloro-N-(4-fluorophenyl)-1,3,5-triazin-2-amine
Molecular Weight (g/mol): 259.07
Molecular Formula: C9H5Cl2FN4
InChi Key: GVFNRSSFMJILLU-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC2=NC(Cl)=NC(Cl)=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results