accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131488-83-0

IUPAC Name: 2-[3-hydroxy-2-(pent-2-en-1-yl)cyclopentyl]acetic acid
Synonyms: [3-hydroxy-2-(pent-2-en-1-yl)cyclopentyl]acetic acid
Molecular Weight (g/mol): 212.29
Molecular Formula: C12H20O3
InChi Key: LYSGIJUGUGJIPS-UHFFFAOYNA-N
SMILES: CCC=CCC1C(O)CCC1CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results