accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1315-05-5

SeSbSeSbSe

CAS RN 1315-05-5

IUPAC Name: bicyclo[1.1.1]distibaselenane
Synonyms: antimony triselenide
Molecular Weight (g/mol): 480.43
Molecular Formula: Sb2Se3
InChi Key: GNZJTRGEKSBAAS-UHFFFAOYSA-N
SMILES: [Se]1[Sb]2[Se][Sb]1[Se]2
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)