accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1315277-53-2

IUPAC Name: 2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 266.57
Molecular Formula: C14H20BClO2
InChi Key: WZAJMSPJXKZSTL-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC2=CC(Cl)=CC=C2)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results