accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNH3CCl

CAS RN 1315281-29-8

IUPAC Name: 2-chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
Synonyms: 2-chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
Molecular Weight (g/mol): 242.51
Molecular Formula: C10H16BClN2O2
InChi Key: MOQXZCYTVMKINQ-UHFFFAOYSA-N
SMILES: CN1C(Cl)=NC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results