accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOOONHNHH3CCH3CH3H3C

CAS RN 1315449-94-5

IUPAC Name: 6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylhexanoic acid
Synonyms: 6-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylhexanoic acid
Molecular Weight (g/mol): 482.58
Molecular Formula: C27H34N2O6
InChi Key: MYQXEVZHWDILHG-HHHXNRCGSA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)