accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOOHCl

CAS RN 13159-52-9

IUPAC Name: 3-chloro-2-hydroxypropyl 2-methylprop-2-enoate
Synonyms: 3-chloro-2-hydroxypropyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 178.61
Molecular Formula: C7H11ClO3
InChi Key: DDKMFQGAZVMXQV-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OCC(O)CCl
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results