accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeN

CAS RN 1316-91-2

IUPAC Name: Ferroceneacetonitrile
Synonyms: —
Molecular Weight (g/mol): 225.07
Molecular Formula: C12H11FeN
InChi Key: —
SMILES: [Fe].c1cccc1.N#CCc1cccc1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results