CAS RN 1316-98-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
tert-Butylferrocene, 98%, Thermo Scientific Chemicals
CAS: 1316-98-9 MDL Number: MFCD00058729
2
tert-Butylferrocene 97.0+%, TCI America™
CAS: 1316-98-9 Molecular Formula: C14H18Fe MDL Number: MFCD00058729