accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3CH3H3CHFHFHF

CAS RN 131600-43-6

IUPAC Name: ethylbis(propan-2-yl)amine trihydrofluoride
Synonyms: N,N-diisopropylethylamine trihydrofluoride
Molecular Weight (g/mol): 189.27
Molecular Formula: C8H22F3N
InChi Key: AJRRXKJZYYBJPY-UHFFFAOYSA-N
SMILES: F.F.F.CCN(C(C)C)C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)