accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CHNONHCH3CH3H3COOCH3(?)

CAS RN 13161-28-9

IUPAC Name: 9-[2-(ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethyl-10λ⁴-xanthen-10-ylium chloride
Synonyms: rhodamine 6G
Molecular Weight (g/mol): 479.02
Molecular Formula: C28H31ClN2O3
InChi Key: XFKVYXCRNATCOO-UHFFFAOYSA-M
SMILES: [Cl-].CCNC1=CC2=[O+]C3=CC(NCC)=C(C)C=C3C(C3=CC=CC=C3C(=O)OCC)=C2C=C1C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results